Learn More
Cayman Chemical Arachidonic Acid ethyl ester
SDP
Catalog No. 502889531
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure
Catalog No. 50-288-9531 Supplier Cayman Chemical Supplier No. 1000820025 MG
Add to Cart
Add to Cart
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure