Learn More
Cayman Chemical Arachidonic Acid ethyl ester
SDP
Catalog No. 502890923
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure
Catalog No. 50-289-0923 Supplier Cayman Chemical Supplier No. 10008200100 MG
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure