Learn More
Cayman Chemical Arachidonic Acid Leelamide
SDP
Supplier:  Cayman Chemical 1000861725 MG
Encompass_Preferred
Arachidonic acid leelamide is the arachidonic amide analog of leelamine. Leelamine has weak affinity for the human CB1 and CB2 receptors, exhibiting 20% displacement of [3H]-CP55940 at a concentration of 10 µM.{14127} Leelamine inhibits PDK with an IC50 value of 9.5 µM.{14105} Derivatives of leelamine exhibit anti-inflammatory activity and show moderate inhibition of PLA2 activity from a variety of sources.{14107} There are no published studies of the pharmacological properties of arachidonic acid leelamide.
Catalog No. 50-288-9419