Learn More
Cayman Chemical Arachidonic Acid Leelamide
SDP
Catalog No. 502889418
Encompass_Preferred
Arachidonic acid leelamide is the arachidonic amide analog of leelamine. Leelamine has weak affinity for the human CB1 and CB2 receptors, exhibiting 20% displacement of [3H]-CP55940 at a concentration of 10 µM.{14127} Leelamine inhibits PDK with an IC50 value of 9.5 µM.{14105} Derivatives of leelamine exhibit anti-inflammatory activity and show moderate inhibition of PLA2 activity from a variety of sources.{14107} There are no published studies of the pharmacological properties of arachidonic acid leelamide.
Catalog No. 50-288-9418 Supplier Cayman Chemical Supplier No. 1000861710 MG
Add to Cart
Add to Cart
Arachidonic acid leelamide is the arachidonic amide analog of leelamine. Leelamine has weak affinity for the human CB1 and CB2 receptors, exhibiting 20% displacement of [3H]-CP55940 at a concentration of 10 µM.{14127} Leelamine inhibits PDK with an IC50 value of 9.5 µM.{14105} Derivatives of leelamine exhibit anti-inflammatory activity and show moderate inhibition of PLA2 activity from a variety of sources.{14107} There are no published studies of the pharmacological properties of arachidonic acid leelamide.