Learn More
Cayman Chemical Arachidonic Acid MaxSpec Standard, Purity: ≥95%, CAS: 506-32-1, 1 mg
SDP
Catalog No. 502890940
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-289-0940 Supplier Cayman Chemical Supplier No. 100072681 MG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications