Learn More
Cayman Chemical Arachidonic Acid-d11, Purity: ≥99% deuterated forms (d1-d11), CAS: 2692624-11-4, 1 mg
SDP
Catalog No. 502889827
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. 50-288-9827 Supplier Cayman Chemical Supplier No. 100067581 MG
An internal standard for the quantification of arachidonic acid by GC- or LC-MS