Learn More
Cayman Chemical ArachIdonIc AcIdpRoxID fr 50mg
SDP
Catalog No. 502895115
Encompass_Preferred
An ω-6 polyunsaturated fatty acid and the precursor of eicosanoids; keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals
Catalog No. 50-289-5115 Supplier Cayman Chemical Supplier No. 90010.150 MG
An ω-6 polyunsaturated fatty acid and the precursor of eicosanoids; keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals