Learn More
Enzo Life Sciences Arachidonoyl-AMC (5 mg)
SDP
Catalog No. 502012189
Encompass_Preferred
Fluorogenic substrate for fatty acid amide hydrolase (FAAH). This substrate is the basis for a simple, selective and sensitive assay for FAAH which is amenable to high-throughput screening. Purity: ≥99%. Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-2189 Supplier Enzo Life Sciences Supplier No. BMLST5040005
Fluorogenic substrate for fatty acid amide hydrolase (FAAH). This substrate is the basis for a simple, selective and sensitive assay for FAAH which is amenable to high-throughput screening. Purity: ≥99%. Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: -80°C.