Learn More
Cayman Chemical Arachidonoyl Ethanolamide Max
SDP
Catalog No. 502889218
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
This item is not returnable. View return policy
Catalog No. 50-288-9218 Supplier Cayman Chemical Supplier No. 10007270100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications