Learn More
Cayman Chemical Arachidonoyl Ethanolamide-d4, 946524-40-9, 1 mg
SDP
Supplier:  Cayman Chemical 100111781
Encompass_Preferred
An internal standard for the quantification of AEA by GC- or LC-MS
Catalog No. NC1315072