Learn More
Cayman Chemical ArachidonoylNmethyl amide
SDP
Catalog No. 502887928
Encompass_Preferred
An analog of AEA that binds to the human CB1 receptor with a Ki value of 60 nM; inhibits rat glial gap junction cell-cell communication at a concentration of 50 µM
Catalog No. 50-288-7928 Supplier Cayman Chemical Supplier No. 100072945 MG
An analog of AEA that binds to the human CB1 receptor with a Ki value of 60 nM; inhibits rat glial gap junction cell-cell communication at a concentration of 50 µM