Learn More
Cayman Chemical ArachIdonyl 2CLOroethylam 50mg
SDP
Catalog No. 502894720
Encompass_Preferred
A potent and selective CB1 receptor agonist (Kis = 1.4 and >2,000 nM for CB1 and CB2 receptors, respectively); induces hypothermia in mice with the same efficacy as AEA in spite of its higher affinity for the CB1 receptor and thus is a possible substrate for FAAH
Catalog No. 50-289-4720 Supplier Cayman Chemical Supplier No. 9105450 MG
A potent and selective CB1 receptor agonist (Kis = 1.4 and >2,000 nM for CB1 and CB2 receptors, respectively); induces hypothermia in mice with the same efficacy as AEA in spite of its higher affinity for the CB1 receptor and thus is a possible substrate for FAAH