Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arachidonyl alcohol | 13487-46-2 | 98.0% | C20H34O | 10MG
SDP

Catalog No. 5000259739
Encompass_Preferred

Arachidonyl alcohol is a long-chain primary fatty alcohol (C20H34O) used as a substrate in the synthesis of ether lipids and related biochemical studies. Supplied at high purity for research use only, it is a colorless to light-yellow liquid with a density around 0.878 g/cm3. Solubility and storage characteristics support its use in both in vitro and formulated in vivo applications.

  • High purity: 98.0%.
  • Chemical formula: C20H34O.
  • Appearance: colorless to light yellow liquid; density ≈0.878 g/cm3.
  • Solubility: soluble in DMSO at 100 mg/mL; in vivo formulations achievable with co-solvents.
  • Storage: stable at -20°C for long-term storage; in solvent stability at -80°C for months.
  • Application: suitable substrate for ether-lipid synthesis and biochemical assays.
  • Intended for research use only; not for human use.

Catalog No. 50-002-59739 Supplier Medchemexpress LLC Supplier No. HY13580110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arachidonyl alcohol is a long-chain primary fatty alcohol (C20H34O) used as a substrate in the synthesis of ether lipids and related biochemical studies. Supplied at high purity for research use only, it is a colorless to light-yellow liquid with a density around 0.878 g/cm3. Solubility and storage characteristics support its use in both in vitro and formulated in vivo applications.

  • High purity: 98.0%.
  • Chemical formula: C20H34O.
  • Appearance: colorless to light yellow liquid; density ≈0.878 g/cm3.
  • Solubility: soluble in DMSO at 100 mg/mL; in vivo formulations achievable with co-solvents.
  • Storage: stable at -20°C for long-term storage; in solvent stability at -80°C for months.
  • Application: suitable substrate for ether-lipid synthesis and biochemical assays.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.