Learn More
Cayman Chemical Arachidonoyl Chloride, Purity: ≥95%, CAS: 57303-04-5, 10 mg
SDP
Catalog No. 502968329
Encompass_Preferred
A derivative of arachidonic acid; has been used as an intermediate in the synthesis of arachidonic acid derivatives
Catalog No. 50-296-8329 Supplier Cayman Chemical Supplier No. 1807810 MG
A derivative of arachidonic acid; has been used as an intermediate in the synthesis of arachidonic acid derivatives