Learn More
Cayman Chemical Arachidonoyl Chloride, Purity: ≥95%, CAS: 57303-04-5, 5 mg
SDP
Catalog No. 502967764
Encompass_Preferred
A derivative of arachidonic acid; has been used as an intermediate in the synthesis of arachidonic acid derivatives
Catalog No. 50-296-7764 Supplier Cayman Chemical Supplier No. 180785 MG
A derivative of arachidonic acid; has been used as an intermediate in the synthesis of arachidonic acid derivatives