Learn More
Cayman Chemical ArachIdonyl EthnolamIDd8 100ug
SDP
Catalog No. 502917122
Encompass_Preferred
An internal standard for the quantification of AEA by GC- or LC-MS.
Catalog No. 50-291-7122 Supplier Cayman Chemical Supplier No. 390050100 UG
An internal standard for the quantification of AEA by GC- or LC-MS.