Learn More
Cayman Chemical Arachidonoyl Ethanolamide, Purity: ≥98%, CAS: 94421-68-8, 50 mg
SDP
Catalog No. 502895387
Encompass_Preferred
An endogenous CB neurotransmitter that has Ki values ranging from 61 to 543 nM for CB1 receptors and from 279 to 1,940 nM for CB2 receptors
Catalog No. 50-289-5387 Supplier Cayman Chemical Supplier No. 9005050 MG
An endogenous CB neurotransmitter that has Ki values ranging from 61 to 543 nM for CB1 receptors and from 279 to 1,940 nM for CB2 receptors