Learn More
Cayman Chemical ArachIdonyl GlycIn 100mg
SDP
Catalog No. 502895388
Encompass_Preferred
Produced endogenously via oxidation of AEA, or by transacylation of arachidonoyl CoA; has analgesic activities in whole animal experiments, yet is a very poor ligand for the CB1 receptor
Catalog No. 50-289-5388 Supplier Cayman Chemical Supplier No. 90051100 MG
Produced endogenously via oxidation of AEA, or by transacylation of arachidonoyl CoA; has analgesic activities in whole animal experiments, yet is a very poor ligand for the CB1 receptor