Learn More
Cayman Chemical ArachIdonyl GlycIn 10mg
SDP
Catalog No. 502896152
Encompass_Preferred
Produced endogenously via oxidation of AEA, or by transacylation of arachidonoyl CoA; has analgesic activities in whole animal experiments, yet is a very poor ligand for the CB1 receptor
Catalog No. 50-289-6152 Supplier Cayman Chemical Supplier No. 9005110 MG
Produced endogenously via oxidation of AEA, or by transacylation of arachidonoyl CoA; has analgesic activities in whole animal experiments, yet is a very poor ligand for the CB1 receptor