Learn More
Cayman Chemical Arachidonoyl Serinol, Purity: ≥98%, CAS: 183718-70-9, 100 mg
SDP
Catalog No. 502893782
Encompass_Preferred
A 2-AG analog with increased stability; approximately a log less potent as a CB1 receptor agonist than 2-AG
Catalog No. 50-289-3782 Supplier Cayman Chemical Supplier No. 62170100 MG
A 2-AG analog with increased stability; approximately a log less potent as a CB1 receptor agonist than 2-AG