Learn More
Cayman Chemical Arachidonoyl Serotonin, Purity: ≥98%, CAS: 187947-37-1, 50 mg
SDP
Catalog No. 502894493
Encompass_Preferred
An inhibitor of FAAH attenuating the FAAH activity from mouse neuroblastoma cells with an IC50 value of 12 µM; alters both the Km and the Vmax of FAAH, indicating that it is a very tight binding, competitive inhibitor; does not inhibit cPLA2 and is essentially devoid of cannabimimetic activity
Catalog No. 50-289-4493 Supplier Cayman Chemical Supplier No. 7066550 MG
An inhibitor of FAAH attenuating the FAAH activity from mouse neuroblastoma cells with an IC50 value of 12 µM; alters both the Km and the Vmax of FAAH, indicating that it is a very tight binding, competitive inhibitor; does not inhibit cPLA2 and is essentially devoid of cannabimimetic activity