Learn More
Cayman Chemical Arachidonoyl thio-PC, Purity: ≥98%, CAS: 146797-82-2, 25 mg
SDP
Catalog No. 502896750
Encompass_Preferred
A chromogenic substrate for many PLA2s including sPLA2, cPLA2, and iPLA2
Catalog No. 50-289-6750 Supplier Cayman Chemical Supplier No. 6224025 MG
A chromogenic substrate for many PLA2s including sPLA2, cPLA2, and iPLA2