Learn More
Cayman Chemical ArachIdonyl2Fluoroethylam 10mg
SDP
Catalog No. 502895795
Encompass_Preferred
An analog of AEA in which the alcohol of the ethanolamide group has been replaced with a fluorine atom, which increases the binding affinity and selectivity for the CB1 receptor; in vivo activity is enhanced much less than the binding affinity, because it is rapidly hydrolyzed by FAAH
Catalog No. 50-289-5795 Supplier Cayman Chemical Supplier No. 9005410 MG
An analog of AEA in which the alcohol of the ethanolamide group has been replaced with a fluorine atom, which increases the binding affinity and selectivity for the CB1 receptor; in vivo activity is enhanced much less than the binding affinity, because it is rapidly hydrolyzed by FAAH