Learn More
Cayman Chemical Arachidonoyl-2'-Fluoroethylamide, Purity: ≥98%, CAS: 166100-37-4, 50 mg
SDP
Catalog No. 502895390
Encompass_Preferred
An analog of AEA in which the alcohol of the ethanolamide group has been replaced with a fluorine atom, which increases the binding affinity and selectivity for the CB1 receptor; in vivo activity is enhanced much less than the binding affinity, because it is rapidly hydrolyzed by FAAH
Catalog No. 50-289-5390 Supplier Cayman Chemical Supplier No. 9005450 MG
An analog of AEA in which the alcohol of the ethanolamide group has been replaced with a fluorine atom, which increases the binding affinity and selectivity for the CB1 receptor; in vivo activity is enhanced much less than the binding affinity, because it is rapidly hydrolyzed by FAAH