Learn More
Bioworld ARACHIS HYPOGAEAPEANUT) -
Catalog No. 50198728
Encompass
ARACHIS HYPOGAEA (PEANUT) - LECTIN: Arachis hypogaea (peanut) Lectin isolated from Arachis hypogaea (peanut). Specifications: Primary sugar specificity: Galactose; Preferential binding activity: galactosyl (beta-1,3) N-acetylgl...
Catalog No. 50-198-728 Supplier Bioworld Supplier No. 215100002
ARACHIS HYPOGAEA (PEANUT) - LECTIN: Arachis hypogaea (peanut) Lectin isolated from Arachis hypogaea (peanut). Specifications: Primary sugar specificity: Galactose; Preferential binding activity: galactosyl (beta-1,3) N-acetylgl...