Learn More
Bioworld ARACHIS HYPOGAEAPEANUT AG
Supplier:  Bioworld 201200002
Encompass
ARACHIS HYPOGAEA (PEANUT AGGLUTININ) SEPAROPORE 4B-CL: Peanut agglutinin (PNA, Peanut Agglutinin) is a 110,000 molecular weight lectin composed of four identical subunits of approximately 27,000 daltons each. PNA binds preferentially to ...
Catalog No. 50-197-514