Learn More
Bioworld ARACHIS HYPOGAEAPEANUT AG
Catalog No. 50197513
Encompass
ARACHIS HYPOGAEA (PEANUT AGGLUTININ) SEPAROPORE 4B-CL: Peanut agglutinin (PNA, Peanut Agglutinin) is a 110,000 molecular weight lectin composed of four identical subunits of approximately 27,000 daltons each. PNA binds preferentially to ...
Catalog No. 50-197-513 Supplier Bioworld Supplier No. 201200001
ARACHIS HYPOGAEA (PEANUT AGGLUTININ) SEPAROPORE 4B-CL: Peanut agglutinin (PNA, Peanut Agglutinin) is a 110,000 molecular weight lectin composed of four identical subunits of approximately 27,000 daltons each. PNA binds preferentially to ...