Learn More
Bioworld Arachis hypogaea (Peanut) Lectin (PNA) - MicroPlate(s)s
Supplier:  Bioworld 201400081
Encompass
High quality Arachis hypogaea (Peanut) Lectin coated microplates.Technical Specifications:Ligand Source: Arachis hypogaeaMolecular Weight:110 kDaCarbohydrate Specificity:Galβ1,3GalNAcInhibiting/Eluting Sugar:GalactoseCarbohydrate for Elution: 0.2M GalactoseBlood Group Specificity:T-antigenForm:96 Well MicroplateIsoelectric (pI): 5.5-6.5Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-197-537