Learn More
ATCC Arachnia propionica; Strain: F0700
Catalog No. 502384565
Encompass
Arachnia propionica strain F0700 was isolated in 2017 from a human in the United States. This bacterial strain is propagated under microaerophilic conditions at 37°C.
Catalog No. 50-238-4565 Supplier ATCC Supplier No. BAA3130
Add to Cart
Add to Cart
Arachnia propionica strain F0700 was isolated in 2017 from a human in the United States. This bacterial strain is propagated under microaerophilic conditions at 37°C.