Learn More
Aviva Systems Biology Corp ARAF Antibody - middle region (ARP54592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARAF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARAF. Alternate names: A-RAF, ARAF1, PKS2, RAFA1ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-61251