Learn More
Electron Microscopy Sciences Araldite 502 Kit
SDP
Catalog No. 50980379
Encompass_Preferred
Luft's Formula. Polymerization takes place overnight so blocks can be sectioned the next day. Reference: J. Biochem. Biophy. Cytol. 9, 409 (1961).
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA. BDMA is less viscous, has a longer shelf life, and offers better penetration. When using BDMA in place of DMP-30 you must vary the amount: DMP-30: 1.5-2%; BDMA: 2.5-3%.
Catalog No. 50-980-379 Supplier Electron Microscopy Sciences Supplier No. 13900
Luft's Formula. Polymerization takes place overnight so blocks can be sectioned the next day. Reference: J. Biochem. Biophy. Cytol. 9, 409 (1961).
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA. BDMA is less viscous, has a longer shelf life, and offers better penetration. When using BDMA in place of DMP-30 you must vary the amount: DMP-30: 1.5-2%; BDMA: 2.5-3%.