Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Araldite 502 Resin 450 ML
SDP

Catalog No. 50980186
Encompass_Preferred

W.P.E. number is included on each bottle.
Specific Gravity: 1.13
CAS: 84-74-2

Catalog No. 50-980-186 Supplier Electron Microscopy Sciences Supplier No. 10900
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

W.P.E. number is included on each bottle.
Specific Gravity: 1.13
CAS: 84-74-2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.