Learn More
Electron Microscopy Sciences Araldite 6005 Kit
SDP
Catalog No. 50980380
Encompass_Preferred
Also known as American Araldite. The blocks are slightly harder than those produced by Araldite 502.
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration.
Catalog No. 50-980-380 Supplier Electron Microscopy Sciences Supplier No. 13920
Also known as American Araldite. The blocks are slightly harder than those produced by Araldite 502.
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration.