Learn More
Electron Microscopy Sciences Araldite Embedding Kit
SDP
Supplier:  Electron Microscopy Sciences 13940
Encompass_Preferred
Mollenhauer Epon-Araldite Formula. For hard blocks and high image contrast. Blocks are easily sectioned.
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA. BDMA is less viscous, has a longer shelf life, and offers better penetration. When using BDMA in place of DMP-30 you must vary the amount: DMP-30: 1.5-2%; BDMA: 2.5-3%.
Catalog No. 50-980-381