Learn More
Wells Lamont™ Aramid Fiber Sleeves
GSA_VA
Catalog No. 19037306 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Length (English):
14 in.
16 in.
18 in.
Style:
Sleeve with Thumb Hole
Standard Sleeve
Construction:
2-ply
Aramid Fiber with one track
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Style Construction
19-037-306 18 in. Sleeve with Thumb Hole 2-ply
19-013437 14 in. Standard Sleeve 2-ply
19-053941 18 in. Sleeve with Thumb Hole Aramid Fiber with one track
19-013439 16 in. Standard Sleeve 2-ply
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-037-306 Supplier Wells Lamont™ Supplier No. SK18H
  • $169.30
    $96.65 / Dozen of 12
    Save $72.65
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
These 2 ply Aramid Fiber Sleeves are an ideal solution in providing arm protection and comfort. ANSI A3 rating provides great cut protection, and the material made from para-aramid yarns adds safety with flame-resistant properties. Offer excellent protection against cutting and burn hazards.
  • 2-ply knit sleeve
  • Flame resistant
  • Available with or without a thumb hole
  • ANSI Level 2 Heat Rating
  • ANSI Level 4 Flame Rating
  • ANSI Cut Level A3
  • Made in the USA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Knit Sleeve
Construction 2-ply
Material Aramid Fiber
Color Yellow
Style Sleeve with Thumb Hole
Disposable Reusable
Cleaning Procedure Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution.
Length (English) 18 in.
Length (Metric) 45.7 cm