Learn More
KETCHUM MFG CO INC Aramis Laboratory Animal Mircotattoo System
Catalog No. NC3847505
Encompass
The Aramis Laboratory Animal Microtattoo System is a quick, efficient and affordable method of permanently identifying laboratory animals (particularly mice and rats as early as 1 day old).Designed and tested by laboratory animal technicians, the system utilizes a hypodermic needle at one end of a stainless steel forcep and a small metal cup containing Ketchum Green Tattoo Paste at the other side of the forcep. By squeezing the forcep together, the needle penetrates the toe/foot pad, ear or tail of the animal and injects a deposit of green paste.The Aramis Microtattoo System includes: 2 Hypodermic Needles1 Plated Steel Microtattoo Forceps1 oz. of Green Tattoo Paste1 Aluminum Planchette1 Storage Case
Catalog No. NC3847505 Supplier KETCHUM MFG CO INC Supplier No. 623
The Aramis Laboratory Animal Microtattoo System is a quick, efficient and affordable method of permanently identifying laboratory animals (particularly mice and rats as early as 1 day old).Designed and tested by laboratory animal technicians, the system utilizes a hypodermic needle at one end of a stainless steel forcep and a small metal cup containing Ketchum Green Tattoo Paste at the other side of the forcep. By squeezing the forcep together, the needle penetrates the toe/foot pad, ear or tail of the animal and injects a deposit of green paste.The Aramis Microtattoo System includes: 2 Hypodermic Needles1 Plated Steel Microtattoo Forceps1 oz. of Green Tattoo Paste1 Aluminum Planchette1 Storage Case