Learn More
Braintree Scientific Aramis Micro-tattoo System
SDP
Catalog No. NC9080035
Encompass
Micro-tattooing forceps with disposable hypodermic needle. Individual identification is essential for maintenance of health/study records and management of colonies of laboratory animals. This instrument is in compliance with regulatory agencies and research protocol requirements for care and use of laboratory animals. Advantages: Permanent mark is easily readable on pigmented or albino animals, Humane, Rapid and easy to apply, Almost infinite ID numbers, Sterile and disposable hypodermic needles prevent spread of potentially infectious disease, Inexpensive. **Sold for research purposes only and not for human use**
Catalog No. NC9080035 Supplier Braintree Scientific Supplier No. MTKARAMIS
Micro-tattooing forceps with disposable hypodermic needle. Individual identification is essential for maintenance of health/study records and management of colonies of laboratory animals. This instrument is in compliance with regulatory agencies and research protocol requirements for care and use of laboratory animals. Advantages: Permanent mark is easily readable on pigmented or albino animals, Humane, Rapid and easy to apply, Almost infinite ID numbers, Sterile and disposable hypodermic needles prevent spread of potentially infectious disease, Inexpensive. **Sold for research purposes only and not for human use**