Learn More
AdipoGen Aranciamycin
SDP
Supplier:  AdipoGen BVT0364C500
Encompass_Preferred
Chemical. CAS 72389-06-1. Formula C27H28O12. MW 544.5. Isolated from Streptomyces echinatus. Antibiotic. Similar to steffimycin. Active against Gram-positive bacteria. Clostridium histolyticum collagenase inhibitor. Antitumor compound.
Catalog No. 50-256-1972