Learn More
AdipoGen Aranorosin (250 µg)
SDP
Supplier:  AdipoGen AGCN20114C250
Encompass_Preferred
Chemical. CAS: 117184-53-9. Formula: C23H33NO6. MW: 419.5. Isolated from Gymnascella sp. Antibiotic. Antibacterial. Antifungal. Anticancer compound. Inhibits anti-apoptotic functions regulated by Bcl-2. Circumvents arbekacin (ABK)-resistance in MRSA by inhibiting the bifunctional enzyme AAC(6')/APH(2'').
Catalog No. 50-205-3478