Learn More
AdipoGen Aranorosinol B (1 mg)
SDP
Supplier:  AdipoGen AGCN20494M001
Encompass_Preferred
Chemical. CAS: 145147-05-3. Formula: C26H39NO7. MW: 477.6. Isolated from Phoma sp. Antibiotic. Moderate antibacterial and antifungal agent. Inhibits the autophosphorylation of histidine kinase WalK/YycG. The WalK/WalR (aka YycG/YycF) two-component system is indispensable in the signal transduction pathway for the cell-wall metabolism/synthesis.
Catalog No. 50-205-3678