Learn More
Apexbio Technology LLC Arbidol HCl 131707-23-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096219
Encompass_Preferred
Arbidol HCl (131707-23-8) is a small-molecule inhibitor designed to interfere with the fusion between viral envelope proteins and host cell membranes thereby preventing viral internalization into target cells Arbidol HCl exerts its biological activity primarily through inhibition of viral entry steps In cellular assays Arbidol HCl demonstrates inhibitory activity with IC50 values typically ranging from approximately 5 to 20 M depending on viral strain and experimental conditions Based on these pharmacological properties Arbidol HCl holds research potential in studies of virus-host interactions antiviral drug discovery and investigations of enveloped viral infection mechanisms
Catalog No. 50-000-96219 Supplier Apexbio Technology LLC Supplier No. A83625.1
Arbidol HCl (131707-23-8) is a small-molecule inhibitor designed to interfere with the fusion between viral envelope proteins and host cell membranes thereby preventing viral internalization into target cells Arbidol HCl exerts its biological activity primarily through inhibition of viral entry steps In cellular assays Arbidol HCl demonstrates inhibitory activity with IC50 values typically ranging from approximately 5 to 20 M depending on viral strain and experimental conditions Based on these pharmacological properties Arbidol HCl holds research potential in studies of virus-host interactions antiviral drug discovery and investigations of enveloped viral infection mechanisms