Learn More
Buehler ARBOR WAFERIN G BLADE 4IN
Supplier:  Buehler 114244
Encompass
1/2 in. (12.7mm) Arbor Wafering Blades, Diameter:4 in. (102mm), Thickness:0.012 in. (0.3mm), Series 15 High Concentration Diamond, for routine use, metal matrix composites, PC boards, bone, titanium, thermal spray coatings
Catalog No. NC9236967