Learn More
Buehler ARBOR WAFERIN G BLADE 5IN
Supplier:  Buehler 114245
Encompass
1/2 in. (12.7mm) Arbor Wafering Blades, Dia:5 in. (127mm), Thick:0.015 in. (0.4mm), Use with Saws:All, Series 15 High Concentration Diamond, for routine use, metal matrix composites, PC boards, bone, titanium, thermal spray coatings
Catalog No. NC9261375