Learn More
Buehler ARBOR WAFERING BLADE
Catalog No. NC9417578
Encompass
Series 15LC Diamond Wafering Blade, Diameter: 5 in. , Thickness: 0.015 in.
Catalog No. NC9417578 Supplier Buehler Supplier No. 114255
Series 15LC Diamond Wafering Blade, Diameter: 5 in. , Thickness: 0.015 in.