Learn More
AdipoGen Arbutin (10 g)
SDP
Supplier:  AdipoGen CDXA0402G010
Encompass_Preferred
Chemical. CAS: 497-76-7. Formula: C12H16O7. MW: 272.25. Arbutin is a glycosylated hydroquinone used in traditional Chinese medicine (TCM) that has been found in Arctostaphylos plants and has diverse biological activities, including tyrosinase inhibitory, antioxidant, anti-tumor and anti-inflammatory properties. Arbutin is a competitive inhibitor of tyrosinase in melanocytes (suppressing production of melanin) and therefore used as a skin-lightening agent and in the treatment of hyperpigmentation disorders.
Catalog No. 50-205-4267