Learn More
Medchemexpress LLC α-Arbutin | 84380-01-8 | 1 ML
SDP
Catalog No. 5000421200
Encompass_Preferred
α-Arbutin is a tyrosinase inhibitor used as an effective skin whitener. It shows promise for research into various conditions such as hyperpigmentation disorders, certain cancers, central nervous system disorders, osteoporosis, and diabetes.
  • Significantly inhibits melanin synthesis at noncytotoxic concentrations
  • Reduces cellular tyrosinase activity
  • Decreases melanin synthesis in a three-dimensional cultured skin model
Catalog No. 50-004-21200 Supplier Medchemexpress LLC Supplier No. HYN300210MM/1ML
α-Arbutin is a tyrosinase inhibitor used as an effective skin whitener. It shows promise for research into various conditions such as hyperpigmentation disorders, certain cancers, central nervous system disorders, osteoporosis, and diabetes.
  • Significantly inhibits melanin synthesis at noncytotoxic concentrations
  • Reduces cellular tyrosinase activity
  • Decreases melanin synthesis in a three-dimensional cultured skin model