Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arbutin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014326 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4326 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4326 Supplier MilliporeSigma Supelco Supplier No. 6646850MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naturally occurring hydroquinone β-D-glucopyranoside used in the study of cutaneous hyperpigmentation inhibition.

Arbutin is a β-D-glucopyranoside of hydroquinone, that occurs naturally and which can be used in the treatment of various cutaneous hyperpigmentations.Its mode of action basically involves the inhibition of tyrosinase activity of human melanocytes and also suppresses the production of melanin via inhibition of melanin synthetic enzyme. Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arbutin
CAS 497-76-7
Physical Form Neat
Linear Formula C12H16O7
Technique HPLC
MDL Number MFCD00016915
Synonym 4-Hydroxyphenyl-beta-D-glucopyranoside; p-Arbutin; Hydroquinone beta-D-glucopyranoside
Type Analytical Standard
Industry Type Food and Beverage
Molecular Weight (g/mol) 272.25
Quantity 50 mg
Formula Weight 272.25
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.