Learn More
Medchemexpress LLC ARC 239 dihydrochloride | 55974-42-0 | 99.2% | 10 MG
SDP
Catalog No. 5000342420
Encompass_Preferred
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist with pKd values of 5.95, 7.41, and 7.56 at α2A, α2B, and α2C receptors respectively. It binds to CHO cell membranes expressing human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes with pKis of 5.6, 8.4, and 7.08, respectively. The product is intended for research use only and is not sold to patients.
  • Selective α2B/2C adrenoceptor antagonist.
  • Binds to human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes.
  • Purity: 99.20%.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg).
Catalog No. 50-003-42420 Supplier Medchemexpress LLC Supplier No. HY12709A10MG
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist with pKd values of 5.95, 7.41, and 7.56 at α2A, α2B, and α2C receptors respectively. It binds to CHO cell membranes expressing human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes with pKis of 5.6, 8.4, and 7.08, respectively. The product is intended for research use only and is not sold to patients.
  • Selective α2B/2C adrenoceptor antagonist.
  • Binds to human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes.
  • Purity: 99.20%.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg).