Learn More
Medchemexpress LLC ARC 239 dihydrochloride | 55974-42-0 | 99.2% | 25 MG
SDP
Catalog No. 5000339310
Encompass_Preferred
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist, intended for research use only.
  • Selective α2B/2C adrenoceptor antagonist
  • Exhibits pKd values of 5.95 at α2A, 7.41 at α2B, and 7.56 at α2C receptors
  • Binds to CHO cell membranes expressing human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes
  • IC50 & Target includes Alpha-2B adrenergic receptor and 5-HT1A Receptor
Catalog No. 50-003-39310 Supplier Medchemexpress LLC Supplier No. 25MGHY12709A
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist, intended for research use only.
  • Selective α2B/2C adrenoceptor antagonist
  • Exhibits pKd values of 5.95 at α2A, 7.41 at α2B, and 7.56 at α2C receptors
  • Binds to CHO cell membranes expressing human recombinant a2A-, a2B- or a2C-adrenoceptor subtypes
  • IC50 & Target includes Alpha-2B adrenergic receptor and 5-HT1A Receptor