Learn More
Medchemexpress LLC ARC 239 dihydrochloride | 55974-42-0 | 99.2% | 50 MG
SDP
Catalog No. 5000342462
Encompass_Preferred
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist. It binds to CHO cell membranes expressing human recombinant α2A-, α2B-, or α2C-adrenoceptor subtypes. It has pKd values of 5.95, 7.41, and 7.56 at α2A, α2B, and α2C receptors respectively, and pKi values of 5.6, 8.4, and 7.08 for α2A-, α2B- or α2C-adrenoceptor subtypes respectively.
  • Selective α2B/2C adrenoceptor antagonist.
  • High purity of 99.2%.
  • Available in various pack sizes.
Catalog No. 50-003-42462 Supplier Medchemexpress LLC Supplier No. HY12709A50MG
ARC 239 dihydrochloride is a selective α2B/2C adrenoceptor antagonist. It binds to CHO cell membranes expressing human recombinant α2A-, α2B-, or α2C-adrenoceptor subtypes. It has pKd values of 5.95, 7.41, and 7.56 at α2A, α2B, and α2C receptors respectively, and pKi values of 5.6, 8.4, and 7.08 for α2A-, α2B- or α2C-adrenoceptor subtypes respectively.
  • Selective α2B/2C adrenoceptor antagonist.
  • High purity of 99.2%.
  • Available in various pack sizes.